Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
680576
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
-
Chromosome Location
Chr.4: 115060702 - 115160802 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001057850.5 XP_001057850.2 7 - 8 4588 63
XM_006236792.2 XP_006236854.1 7 - 8 4262 63
XM_006236793.2 XP_006236855.1 6 - 7 4205 63
XM_006236794.2 XP_006236856.1 6 - 7 3631 63
XM_008763094.1 XP_008761316.1 8 - 9 3886 63
XM_008763095.1 XP_008761317.1 8 - 9 6633 63

View Full Product Details