Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdh19
Entrez Gene ID
360835
Gene Name
cadherin 19, type 2
Gene Aliases
-
Chromosome Location
Chr.13: 32344954 - 32469909 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009448.1 NP_001009448.1 5 - 6 969 157
XM_006249653.2 XP_006249715.1 6 - 7 1607 157
GenBank mRNA AB121032.1 - 5 - 6 969 157

View Full Product Details