Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prlh
Entrez Gene ID
63850
Gene Name
prolactin releasing hormone
Gene Aliases
Prh, prrp
Chromosome Location
Chr.9: 98111421 - 98114830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF521930.1 - 1 - 2 165 64

View Full Product Details