Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agxt
Entrez Gene ID
24792
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, SPT, Spat
Chromosome Location
Chr.9: 100281339 - 100291292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276706.1 NP_001263635.1 1 - 2 341 65
NM_030656.2 NP_085914.2 1 - 2 341 65
GenBank mRNA BC088133.1 - 1 - 2 173 65
M35270.1 - 1 - 2 341 65
X06357.1 - 1 - 2 279 65

View Full Product Details