Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fer
Entrez Gene ID
301737
Gene Name
fer (fps/fes related) tyrosine kinase
Gene Aliases
Fert2, Flk, Flk_retired
Chromosome Location
Chr.9: 111557802 - 111868583 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106928.1 NP_001100398.1 6 - 7 940 65
XM_006245600.2 XP_006245662.1 11 - 12 1672 65
XM_008767367.1 XP_008765589.1 11 - 12 2448 65
XM_008767369.1 XP_008765591.1 12 - 13 1810 65
XM_008767370.1 XP_008765592.1 11 - 12 2708 65
XM_008767371.1 XP_008765593.1 11 - 12 2445 65
XM_008767372.1 XP_008765594.1 4 - 5 585 65

View Full Product Details