Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agap1
Entrez Gene ID
316611
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
Centg2
Chromosome Location
Chr.9: 96718900 - 97034400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108230.1 NP_001101700.1 6 - 7 640 74
XM_006245396.2 XP_006245458.2 8 - 9 4748 74
XM_006245397.2 XP_006245459.2 8 - 9 4748 74
XM_006245398.2 XP_006245460.2 8 - 9 4747 74
XM_006245400.2 XP_006245462.2 7 - 8 4735 74
XM_006245401.2 XP_006245463.2 8 - 9 4748 74
XM_006245402.2 XP_006245464.2 8 - 9 4748 74
XM_006245403.2 XP_006245465.2 7 - 8 4735 74

View Full Product Details