Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc1
Entrez Gene ID
58920
Gene Name
glypican 1
Gene Aliases
HSPG M12, HSPGM12
Chromosome Location
Chr.9: 99998275 - 100026818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC061572.1 - 2 - 3 538 130
L34067.1 - 2 - 3 373 130

View Full Product Details