The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108235.1 | NP_001101705.1 | 8 - 9 | 1234 | 63 |
GenBank mRNA | BC168863.1 | - | - | 1666 | 63 |