Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqca1
Entrez Gene ID
316616
Gene Name
IQ motif containing with AAA domain 1
Gene Aliases
RGD1305311
Chromosome Location
Chr.9: 97183214 - 97298758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767356.1 XP_008765578.1 13 - 14 1919 76

View Full Product Details