Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd12
Entrez Gene ID
316775
Gene Name
ankyrin repeat domain 12
Gene Aliases
-
Chromosome Location
Chr.9: 113605683 - 113682482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108238.1 NP_001101708.1 11 - 12 6095 79

View Full Product Details