Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf2bp
Entrez Gene ID
499413
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
RGD1565879
Chromosome Location
Chr.20: 10757172 - 10843208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127683.1 NP_001121155.1 3 - 4 305 107
XM_008772826.1 XP_008771048.1 4 - 5 541 107
GenBank mRNA BC158765.1 - 3 - 4 292 107

View Full Product Details