The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013073.1 | NP_001013091.1 | 10 - 11 | 1743 | 64 |
XM_006256185.2 | XP_006256247.1 | 10 - 11 | 1769 | 64 | |
GenBank mRNA | BC087068.1 | - | 10 - 11 | 1743 | 64 |