Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtdr1
Entrez Gene ID
365552
Gene Name
rhabdoid tumor deletion region gene 1
Gene Aliases
-
Chromosome Location
Chr.20: 14577122 - 14653057 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127557.2 NP_001121029.1 1 - 2 249 71
XM_006256340.2 XP_006256402.1 2 - 3 305 71
XM_006256341.2 XP_006256403.1 2 - 3 358 71
GenBank mRNA BC162057.1 - 1 - 2 251 71

View Full Product Details