Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr46
Entrez Gene ID
309628
Gene Name
WD repeat domain 46
Gene Aliases
Bing4
Chromosome Location
Chr.20: 5447860 - 5455632 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212491.2 NP_997656.1 1 - 2 125 70
GenBank mRNA BC085934.1 - 1 - 2 125 70

View Full Product Details