The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008350.1 | NP_001008351.1 | 3 - 4 | 491 | 62 |
XM_006256191.2 | XP_006256253.1 | 4 - 5 | 1321 | 62 | |
XM_006256192.2 | XP_006256254.1 | 4 - 5 | 1122 | 62 | |
XM_008772812.1 | XP_008771034.1 | 4 - 5 | 1105 | 62 | |
GenBank mRNA | BC086421.1 | - | 3 - 4 | 491 | 62 |
FQ231020.1 | - | 2 - 3 | 186 | 62 |