Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a7
Entrez Gene ID
294281
Gene Name
solute carrier family 39 (zinc transporter), member 7
Gene Aliases
Ke4, RT1-Ke4, Rt1Ke4
Chromosome Location
Chr.20: 3819416 - 3822788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008885.2 NP_001008885.1 2 - 3 883 118
NM_001164744.1 NP_001158216.1 3 - 4 624 118
GenBank mRNA BC079141.1 - 2 - 3 883 118
FQ212335.1 - 3 - 4 591 118
FQ212833.1 - 2 - 3 911 118

View Full Product Details