Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jmjd1c
Entrez Gene ID
171120
Gene Name
jumonji domain containing 1C
Gene Aliases
Pr2
Chromosome Location
Chr.20: 22751743 - 22914108 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191719.1 NP_001178648.1 16 - 17 6036 127
XM_006256359.2 XP_006256421.1 16 - 17 6090 127
XM_006256360.2 XP_006256422.1 15 - 16 5886 127
XM_006256361.2 XP_006256423.1 16 - 17 6113 127
XM_006256362.2 XP_006256424.1 16 - 17 6005 127
XM_006256363.1 XP_006256425.1 12 - 13 5447 127
XM_006256364.1 XP_006256426.1 16 - 17 5999 127
XM_006256365.1 XP_006256427.1 11 - 12 5305 127
GenBank mRNA FQ225499.1 - 3 - 4 361 127
FQ230372.1 - 3 - 4 361 127
FQ231799.1 - 3 - 4 361 127

View Full Product Details