The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191719.1 | NP_001178648.1 | 1 - 2 | 163 | 82 |
XM_006256359.2 | XP_006256421.1 | 1 - 2 | 214 | 82 | |
XM_006256360.2 | XP_006256422.1 | 1 - 2 | 214 | 82 |