Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspear
Entrez Gene ID
365546
Gene Name
thrombospondin-type laminin G domain and EAR repeats
Gene Aliases
RGD1563108
Chromosome Location
Chr.20: 11501513 - 11529210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772856.1 XP_008771078.1 8 - 9 1462 94

View Full Product Details