Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd9
Entrez Gene ID
294318
Gene Name
BTB (POZ) domain containing 9
Gene Aliases
-
Chromosome Location
Chr.20: 8899286 - 9253931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256185.2 XP_006256247.1 1 - 2 93 100

View Full Product Details