The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034032.1 | NP_001029204.1 | 4 - 5 | 534 | 58 |
GenBank mRNA | AB062135.1 | - | 4 - 5 | 536 | 58 |
BC100072.1 | - | 4 - 5 | 534 | 58 | |
FQ214972.1 | - | 4 - 5 | 547 | 58 | |
FQ219445.1 | - | 4 - 5 | 547 | 58 |