The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031020.2 | NP_112282.2 | 10 - 11 | 1147 | 114 |
XM_006256142.2 | XP_006256204.1 | 10 - 11 | 859 | 114 | |
GenBank mRNA | AB000783.1 | - | 10 - 11 | 1146 | 114 |
AF346293.1 | - | 10 - 11 | 844 | 114 | |
BC092193.1 | - | 10 - 11 | 1147 | 114 | |
U73142.1 | - | 10 - 11 | 855 | 114 | |
U91847.1 | - | 10 - 11 | 844 | 114 |