Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dlk1
Entrez Gene ID
114587
Gene Name
delta-like 1 homolog (Drosophila)
Gene Aliases
Pref-1, Zog
Chromosome Location
Chr.6: 133576513 - 133583751 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053744.1 NP_446196.1 - 261 87
XM_008764917.1 XP_008763139.1 - 317 87
GenBank mRNA BC167752.1 - - 281 87
D84336.1 - - 261 87
U25680.1 - - 126 87

View Full Product Details