Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a47
Entrez Gene ID
299316
Gene Name
solute carrier family 25, member 47
Gene Aliases
Hdmcp, Slc24a47
Chromosome Location
Chr.6: 132762306 - 132767940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001509.1 NP_001001509.1 1 - 2 72 66
GenBank mRNA AY603424.1 - 1 - 2 72 66
BC089874.1 - 1 - 2 76 66

View Full Product Details