The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107460.1 | NP_001100930.1 | 1 - 2 | 187 | 89 |
XM_008758705.1 | XP_008756927.1 | 1 - 2 | 186 | 89 |