Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galns
Entrez Gene ID
292073
Gene Name
galactosamine (N-acetyl)-6-sulfatase
Gene Aliases
-
Chromosome Location
Chr.19: 55391913 - 55423052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047851.1 NP_001041316.1 2 - 3 248 89

View Full Product Details