Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prmt10
Entrez Gene ID
291947
Gene Name
protein arginine methyltransferase 10 (putative)
Gene Aliases
RGD1306157
Chromosome Location
Chr.19: 34067008 - 34105622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191599.1 NP_001178528.1 8 - 9 1449 80
XM_006255424.2 XP_006255486.1 8 - 9 1485 80

View Full Product Details