Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pard6a
Entrez Gene ID
307799
Gene Name
par-6 family cell polarity regulator alpha
Gene Aliases
Par-6a, Par6a
Chromosome Location
Chr.19: 37668133 - 37670450 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003653.1 NP_001003653.1 1 - 2 98 86
XM_006255490.2 XP_006255552.1 1 - 2 146 86
GenBank mRNA AY682587.1 - 1 - 2 98 86
DQ011518.1 - 1 - 2 63 86

View Full Product Details