Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tat
Entrez Gene ID
24813
Gene Name
tyrosine aminotransferase
Gene Aliases
-
Chromosome Location
Chr.19: 41675639 - 41686195 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012668.2 NP_036800.1 7 - 8 801 74
GenBank mRNA BC089813.1 - 7 - 8 801 74
M18340.1 - 7 - 8 854 74
X02741.1 - 7 - 8 854 74

View Full Product Details