Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkn1
Entrez Gene ID
29355
Gene Name
protein kinase N1
Gene Aliases
PAK-1, Prkcl1
Chromosome Location
Chr.19: 24803524 - 24831500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017175.2 NP_058871.2 9 - 10 1605 121
XM_006255245.2 XP_006255307.1 9 - 10 2645 121
GenBank mRNA BC061836.1 - 9 - 10 1506 121
D26180.1 - 9 - 10 1556 121
FQ222217.1 - 9 - 10 1605 121
L35634.1 - 9 - 10 1448 121

View Full Product Details