Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd7
Entrez Gene ID
307821
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 7
Gene Aliases
-
Chromosome Location
Chr.19: 39639432 - 39646718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107426.1 NP_001100896.1 4 - 5 483 85

View Full Product Details