Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhps
Entrez Gene ID
288923
Gene Name
deoxyhypusine synthase
Gene Aliases
-
Chromosome Location
Chr.19: 26184665 - 26188755 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004207.1 NP_001004207.1 2 - 3 450 125
GenBank mRNA BC079188.1 - 2 - 3 450 125

View Full Product Details