The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100480.1 | NP_001093950.1 | 3 - 4 | 618 | 69 |
XM_006255444.2 | XP_006255506.1 | 4 - 5 | 693 | 69 | |
GenBank mRNA | AF517017.1 | - | 1 - 2 | 159 | 69 |
FQ215613.1 | - | 4 - 5 | 667 | 69 |