Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ptger1
- Entrez Gene ID
-
25637
- Gene Name
- prostaglandin E receptor 1 (subtype EP1)
- Gene Aliases
-
EP1
- Chromosome Location
-
Chr.19: 24799420 - 24803373 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D88752.1 |
- |
1 - 1 |
1939 |
105 |
View Full Product Details