Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam92b
Entrez Gene ID
361423
Gene Name
family with sequence similarity 92, member B
Gene Aliases
RGD1560673
Chromosome Location
Chr.19: 53740743 - 53754734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772654.1 XP_008770876.1 2 - 3 383 82

View Full Product Details