Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam92b
Entrez Gene ID
361423
Gene Name
family with sequence similarity 92, member B
Gene Aliases
RGD1560673
Chromosome Location
Chr.19: 53740743 - 53754734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772654.1 XP_008770876.1 3 - 4 452 65

View Full Product Details