Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtap
Entrez Gene ID
298227
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
Cc1-6
Chromosome Location
Chr.5: 107711077 - 107777530 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047867.1 NP_001041332.1 3 - 4 232 107
XM_006238382.2 XP_006238444.1 2 - 3 281 107
GenBank mRNA AY325240.1 - 3 - 4 232 107

View Full Product Details