Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sar1a
Entrez Gene ID
361842
Gene Name
secretion associated, Ras related GTPase 1A
Gene Aliases
Sara1
Chromosome Location
Chr.20: 31298269 - 31310676 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256451.2 XP_006256513.1 1 - 2 143 137

View Full Product Details