Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
364227
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 115352716 - 115446618 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108864.1 NP_001102334.1 2 - 3 379 110

View Full Product Details