Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shc3
Entrez Gene ID
114858
Gene Name
SHC (Src homology 2 domain containing) transforming protein 3
Gene Aliases
ShcC
Chromosome Location
Chr.17: 13670520 - 13791043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105743.1 NP_001099213.1 8 - 9 1114 143
GenBank mRNA AB001453.1 - 8 - 9 1328 143

View Full Product Details