Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap2a
Entrez Gene ID
306862
Gene Name
transcription factor AP-2 alpha
Gene Aliases
Tcfap2a
Chromosome Location
Chr.17: 24647833 - 24670457 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107345.1 NP_001100815.1 5 - 6 1154 80
XM_008771570.1 XP_008769792.1 5 - 6 891 80

View Full Product Details