Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdbf2
Entrez Gene ID
501153
Gene Name
zinc finger, DBF-type containing 2
Gene Aliases
RGD1560005
Chromosome Location
Chr.9: 70052087 - 70090847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245044.2 XP_006245106.1 3 - 4 273 137
XM_006245045.2 XP_006245107.1 3 - 4 256 137
XM_006245046.2 XP_006245108.1 3 - 4 303 137
XM_008767157.1 XP_008765379.1 4 - 5 575 137
XM_008767158.1 XP_008765380.1 4 - 5 596 137
XM_008767159.1 XP_008765381.1 5 - 6 658 137
XM_576579.7 XP_576579.4 4 - 5 1282 137

View Full Product Details