The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024363.1 | NP_001019534.2 | 3 - 4 | 518 | 96 |
GenBank mRNA | BC083917.1 | - | 3 - 4 | 518 | 96 |