Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xpa
Entrez Gene ID
298074
Gene Name
xeroderma pigmentosum, complementation group A
Gene Aliases
-
Chromosome Location
Chr.5: 61749767 - 61793963 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106656.1 NP_001100126.1 4 - 5 612 88
XM_006238035.2 XP_006238097.1 4 - 5 728 88
XM_006238036.2 XP_006238098.1 4 - 5 1647 88

View Full Product Details