Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clta
Entrez Gene ID
83800
Gene Name
clathrin, light chain A
Gene Aliases
LCA1, LCA2, LCA3
Chromosome Location
Chr.5: 59490363 - 59509139 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031974.1 NP_114180.1 6 - 7 694 96
GenBank mRNA FQ213034.1 - 6 - 7 702 96
M15882.1 - 6 - 7 694 96

View Full Product Details