Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrp11
Entrez Gene ID
292462
Gene Name
low density lipoprotein receptor-related protein 11
Gene Aliases
-
Chromosome Location
Chr.1: 1702696 - 1731210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106217.1 NP_001099687.1 6 - 7 1393 113

View Full Product Details