Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdss2
Entrez Gene ID
365592
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
-
Chromosome Location
Chr.20: 47966511 - 48192747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014249.1 NP_001014271.1 3 - 4 866 153
XM_008773004.1 XP_008771226.1 3 - 4 938 153
GenBank mRNA BC085898.1 - 3 - 4 866 153

View Full Product Details