Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr2e1
Entrez Gene ID
684085
Gene Name
nuclear receptor subfamily 2, group E, member 1
Gene Aliases
TLX
Chromosome Location
Chr.20: 47288137 - 47306318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113197.1 NP_001106668.1 4 - 5 644 60
GenBank mRNA EU316216.1 - 4 - 5 644 60

View Full Product Details