Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nr2e1
Entrez Gene ID
684085
Gene Name
nuclear receptor subfamily 2, group E, member 1
Gene Aliases
TLX
Chromosome Location
Chr.20: 47288137 - 47306318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113197.1 NP_001106668.1 6 - 7 116
GenBank mRNA EU316216.1 - 6 - 7 116

View Full Product Details