The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108655.1 | NP_001102125.1 | 1 - 2 | 231 | 114 |
GenBank mRNA | BC161902.1 | - | 1 - 2 | 220 | 114 |